Merck
Effects of collision energy and vibrational excitation of CH
The Journal of chemical physics (2017-10-23)
Andrea Cernuto, Allan Lopes, Claire Romanzin, Barbara Cunha de Miranda, Daniela Ascenzi, Paolo Tosi, Glauco Tonachini, Andrea Maranzana, Miroslav Polášek, Jan Žabka, Christian Alcaraz
ABSTRACT
The methyl carbocation is ubiquitous in gaseous environments, such as planetary ionospheres, cometary comae, and the interstellar medium, as well as combustion systems and plasma setups for technological applications. Here we report on a joint experimental and theoretical study on the mechanism of the reaction CH