Merck
ADAM12 Is a Novel Regulator of Tumor Angiogenesis via STAT3 Signaling.
Molecular cancer research : MCR (2017-08-03)
Roopali Roy, Adelle Dagher, Catherine Butterfield, Marsha A Moses
ABSTRACT
ADAM12, (
Sigma-Aldrich
MISSION® esiRNA, targeting human ADAM12
Sigma-Aldrich
MISSION® esiRNA, targeting human STAT3
Sigma-Aldrich
MISSION® esiRNA, targeting human EGFR