Merck
  • Carbohydrate-Catalyzed Enantioselective Alkene Diboration: Enhanced Reactivity of 1,2-Bonded Diboron Complexes.
Carbohydrate-Catalyzed Enantioselective Alkene Diboration: Enhanced Reactivity of 1,2-Bonded Diboron Complexes.
Journal of the American Chemical Society (2016-02-09)
Lichao Fang, Lu Yan, Fredrik Haeffner, James P Morken
ABSTRACT
Catalytic enantioselective diboration of alkenes is accomplished with readily available carbohydrate-derived catalysts. Mechanistic experiments suggest the intermediacy of 1,2-bonded diboronates.
Sigma-Aldrich
TBS-DHG Catalyst, ≥95%