- Polyelectrolyte induced controlled assemblies for the backbone of robust and brilliant Raman tags.
Polyelectrolyte induced controlled assemblies for the backbone of robust and brilliant Raman tags.
Near-field and far-field optical properties of plasmonic materials can be tailored by coupling the existing structures. However, fabricating 3D coupled structures in the solution by molecular linkers may suffer from low yield, low stability (particle aggregates), long reaction time, complex surface modification or multiple purification steps. In this report, stable 3D plasmonic core-satellite assemblies (CSA) with a ~1 nm interior gap accompanied by high field enhancement (|E/E