Merck
  • EB1 and EB3 regulate microtubule minus end organization and Golgi morphology.
EB1 and EB3 regulate microtubule minus end organization and Golgi morphology.
The Journal of cell biology (2017-08-18)
Chao Yang, Jingchao Wu, Cecilia de Heus, Ilya Grigoriev, Nalan Liv, Yao Yao, Ihor Smal, Erik Meijering, Judith Klumperman, Robert Z Qi, Anna Akhmanova
ABSTRACT
End-binding proteins (EBs) are the core components of microtubule plus end tracking protein complexes, but it is currently unknown whether they are essential for mammalian microtubule organization. Here, by using CRISPR/Cas9-mediated knockout technology, we generated stable cell lines lacking EB2 and EB3 and the C-terminal partner-binding half of EB1. These cell lines show only mild defects in cell division and microtubule polymerization. However, the length of CAMSAP2-decorated stretches at noncentrosomal microtubule minus ends in these cells is reduced, microtubules are detached from Golgi membranes, and the Golgi complex is more compact. Coorganization of microtubules and Golgi membranes depends on the EB1/EB3-myomegalin complex, which acts as membrane-microtubule tether and counteracts tight clustering of individual Golgi stacks. Disruption of EB1 and EB3 also perturbs cell migration, polarity, and the distribution of focal adhesions. EB1 and EB3 thus affect multiple interphase processes and have a major impact on microtubule minus end organization.
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, clone TUB 2.1, purified from hybridoma cell culture
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder