Merck
  • Antioxidant and anti-inflammatory effects of cauliflower leaf powder-enriched diet against LPS induced toxicity in rabbits.
Antioxidant and anti-inflammatory effects of cauliflower leaf powder-enriched diet against LPS induced toxicity in rabbits.
Food & function (2017-08-24)
Marilena Larocca, Anna Maria Perna, Amalia Simonetti, Emilio Gambacorta, Alessandra Iannuzzi, Angela Perucatti, Rocco Rossano
ABSTRACT
Brassica phytochemicals exert a broad spectrum of health-promoting activities. The aim of this study was to investigate the possible beneficial effects of a cauliflower leaf powder (CLP)-enriched diet to prevent inflammation and oxidative stress resulting from injection of lipopolysaccharide (LPS) into rabbits. Animals (24 rabbits) were randomly divided into two groups and fed with a standard diet (SD) or a standard diet supplemented with a 100 g kg
Sigma-Aldrich
Malondialdehyde tetrabutylammonium salt, ≥96.0% (NT)
Pricing and availability is not currently available.