Merck
  • Towards the determination of sulfonamides in meat samples: A magnetic and mesoporous metal-organic framework as an efficient sorbent for magnetic solid phase extraction combined with high-performance liquid chromatography.
Towards the determination of sulfonamides in meat samples: A magnetic and mesoporous metal-organic framework as an efficient sorbent for magnetic solid phase extraction combined with high-performance liquid chromatography.
Journal of chromatography. A (2017-04-15)
Lian Xia, Lijie Liu, Xiaoxia Lv, Fei Qu, Guoliang Li, Jinmao You
ABSTRACT
A magnetic, mesoporous core/shell structured Fe