Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 24 ga
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with manual holder
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with manual holder
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with manual holder, needle size 24 ga
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Portable Field Sampler, coating PDMS
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly 50/30µm DVB/CAR/PDMS, needle size 23 ga, metal alloy fiber, for use with autosampler, fiber L 1 cm, pkg of 1 × ea
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly 50/30µm DVB/CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 2 cm
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly 50/30µm DVB/CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler, fiber L 1 cm, pk of 3
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, metal alloy fiber, for use with manual holder
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly 50/30µm DVB/CAR/PDMS, needle size 24 ga, StableFlex fiber, for use with autosampler, fiber L 2 cm, pk of 3
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), fused silica fiber, df 65 μm(PDMS/DVB, for use with manual holder, needle size 23 ga
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with manual holder
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly 50/30µm DVB/CAR/PDMS, needle size 24 ga, StableFlex fiber, for use with manual holder
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME-OC Fiber Assembly, Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 75 μm(Coating thickness includes 65 μm coating + 10 μm OC (overcoating), needle size 23 ga, pkg of 3 ea, fiber L 1 cm