Merck
  • FGF signaling refines Wnt gradients to regulate the patterning of taste papillae.
FGF signaling refines Wnt gradients to regulate the patterning of taste papillae.
Development (Cambridge, England) (2017-05-17)
Michaela Prochazkova, Teemu J Häkkinen, Jan Prochazka, Frantisek Spoutil, Andrew H Jheon, Youngwook Ahn, Robb Krumlauf, Jukka Jernvall, Ophir D Klein
ABSTRACT
The patterning of repeated structures is a major theme in developmental biology, and the inter-relationship between spacing and size of such structures is an unresolved issue. Fungiform papillae are repeated epithelial structures that house taste buds on the anterior tongue. Here, we report that FGF signaling is a crucial regulator of fungiform papillae development. We found that mesenchymal FGF10 controls the size of the papillary area, while overall patterning remains unchanged. Our results show that FGF signaling negatively affects the extent of canonical Wnt signaling, which is the main activation pathway during fungiform papillae development; however, this effect does not occur at the level of gene transcription. Rather, our experimental data, together with computational modeling, indicate that FGF10 modulates the range of Wnt effects, likely via induction of
Sigma-Aldrich
SAG dihydrochloride, ≥98% (HPLC)