Merck
  • Synthesis and characterization of Au-NPs supported on carbon nanotubes: Application for the ultrasound assisted removal of radioactive UO
Synthesis and characterization of Au-NPs supported on carbon nanotubes: Application for the ultrasound assisted removal of radioactive UO
Journal of colloid and interface science (2017-05-23)
Mohammad Hassan Omidi, Farshid Nasiri Azad, Mehrorang Ghaedi, Arash Asfaram, Mohammad Hossein Ahmadi Azqhandi, Lobat Tayebi
ABSTRACT
The present study devoted to description of efficient removal of radioactive UO
Sigma-Aldrich
Arsenazo III, calcium-sensitive dye
Pricing and availability is not currently available.