Merck
  • Toxicity of organic compounds from unresolved complex mixtures (UCMs) to primary fish hepatocytes.
Toxicity of organic compounds from unresolved complex mixtures (UCMs) to primary fish hepatocytes.
Aquatic toxicology (Amsterdam, Netherlands) (2017-07-18)
Karina Petersen, Maria T Hultman, Steven J Rowland, Knut Erik Tollefsen
ABSTRACT
Many environmental matrices contaminated with organic pollutants derived from crude oil or degraded petroleum contain mixtures so complex that they are typically unresolved by conventional analytical techniques such as gas chromatography. The resulting chromatographic features have become known as 'humps' or unresolved complex mixtures (UCMs). These UCMs often dominate the organic contaminants of polluted environmental samples: for example, in oil sands produced water up to 150mgL
Sigma-Aldrich
1-Adamantanecarboxylic acid, 99%