Merck
  • Fast and Simple Determination of 3-Aminopiperidine without Derivatization Using High Performance Liquid Chromatography-Charged Aerosol Detector with an Ion-Exchange/Reversed-Phase Mixed-mode Column.
Fast and Simple Determination of 3-Aminopiperidine without Derivatization Using High Performance Liquid Chromatography-Charged Aerosol Detector with an Ion-Exchange/Reversed-Phase Mixed-mode Column.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2017-03-18)
Shubo Dong, Zhengyu Yan, Hanyue Yang, Zhen Long
ABSTRACT
A sensitive non-derivatization method for the determination of the highly polar compound 3-aminopiperidine was developed using a mixed-mode column combined with a charged aerosol detector (CAD). Chromatographic conditions, including the type of detector, separation mode, and mobile phase composition, were optimized to achieve high sensitivity towards and sufficient retention of 3-aminopiperidine. Compared to the precolumn derivatization UV method, the current method showed higher recovery and greater simplicify. High sensitivity (LOQ <2.73 μg mL
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Pricing and availability is not currently available.