- Ultrasensitive and Selective Sensing of Selenium Using Nitrogen-Rich Ligand Interfaced Carbon Quantum Dots.
Ultrasensitive and Selective Sensing of Selenium Using Nitrogen-Rich Ligand Interfaced Carbon Quantum Dots.
ACS applied materials & interfaces (2017-04-01)
Pooja Devi, Anupma Thakur, Shweta Chopra, Navneet Kaur, Praveen Kumar, Narinder Singh, Mahesh Kumar, Sonnada Math Shivaprasad, Manoj K Nayak
PMID28362083
ABSTRACT
This work reports a label-free, ultrasensitive, and selective optical chemosensory system for trace level detection of selenite (SeO