Merck
  • Two alternative pathways for docosahexaenoic acid (DHA, 22:6n-3) biosynthesis are widespread among teleost fish.
Two alternative pathways for docosahexaenoic acid (DHA, 22:6n-3) biosynthesis are widespread among teleost fish.
Scientific reports (2017-06-22)
Angela Oboh, Naoki Kabeya, Greta Carmona-Antoñanzas, L Filipe C Castro, James R Dick, Douglas R Tocher, Oscar Monroig
ABSTRACT
Docosahexaenoic acid (DHA) plays important physiological roles in vertebrates. Studies in rats and rainbow trout confirmed that DHA biosynthesis proceeds through the so-called "Sprecher pathway", a biosynthetic process requiring a Δ6 desaturation of 24:5n-3 to 24:6n-3. Alternatively, some teleosts possess fatty acyl desaturases 2 (Fads2) that enable them to biosynthesis DHA through a more direct route termed the "Δ4 pathway". In order to elucidate the prevalence of both pathways among teleosts, we investigated the Δ6 ability towards C
Sigma-Aldrich
Stearidonic acid, ≥99%