Merck
  • Diadenosine pentaphosphate affects electrical activity in guinea pig atrium via activation of potassium acetylcholine-dependent inward rectifier.
Diadenosine pentaphosphate affects electrical activity in guinea pig atrium via activation of potassium acetylcholine-dependent inward rectifier.
The journal of physiological sciences : JPS (2016-12-13)
Denis V Abramochkin, Viktoria M Karimova, Tatiana S Filatova, Andre Kamkin
ABSTRACT
Diadenosine pentaphosphate (Ap5A) belongs to the family of diadenosine polyphosphates, endogenously produced compounds that affect vascular tone and cardiac performance when released from platelets. The previous findings indicate that Ap5A shortens action potentials (APs) in rat myocardium via activation of purine P2 receptors. The present study demonstrates alternative mechanism of Ap5A electrophysiological effects found in guinea pig myocardium. Ap5A (10
Sigma-Aldrich
Chromanol 293B, ≥98% (HPLC), powder
Sigma-Aldrich
E-4031, ≥98% (HPLC), lyophilized powder
Sigma-Aldrich
8-Cyclopentyl-1,3-dipropylxanthine, solid