Merck
  • LC-MS/MS methods for sulfadimethoxine and ormetoprim analysis in feed and fish fillet and a leaching study for feed after alternative procedures for the incorporation of drugs.
LC-MS/MS methods for sulfadimethoxine and ormetoprim analysis in feed and fish fillet and a leaching study for feed after alternative procedures for the incorporation of drugs.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2016-12-06)
Ana Paula Fais, Rodolfo Scarpino Barboza Franco, Agnaldo Fernando Baldo da Silva, Osvaldo de Freitas, Jonas Augusto Rizzato Paschoal
ABSTRACT
This paper describes the method development for sulfadimethoxine (SDM) and ormetoprim (OMP) quantitation in fish feed and fish fillet employing LC-MS/MS. In order to assess the reliability of the analytical method, valuation was undertaken as recommended by guidelines proposed by the Brazilian Ministry of Agriculture. The calibration curve for the quantification of both drugs in feed showed adequate linearity (r > 0.99), precision (CV < 12%) and trueness ranging from 97% to 100%. The method for the determination of SDM and OMP residues in fish fillet involved a simple sample preparation procedure that had adequate linearity (r > 0.99), precision (CV < 16%) and trueness around 100%, with CCα < 100.2 ng g
Sigma-Aldrich
Orotidine 5′-monophosphate trisodium salt, ≥99% (HPLC), powder