Merck
Baclofen antagonism by 2-hydroxy-saclofen in the cat spinal cord.
Neuroscience letters (1988-09-23)
D R Curtis, B D Gynther, D T Beattie, D I Kerr, R H Prager
ABSTRACT
When administered microelectrophoretically, a sulphonic acid derivative of baclofen, 3-amino-2-(4-chlorophenyl)-2-hydroxy-propylsulphonic acid, reversibly reduced the presynaptic reduction by (-)-baclofen of the monosynaptic excitation of spinal interneurones by impulses in low threshold primary afferent fibres of the cat as well as the postsynaptic depression by (-)-baclofen of the firing of these neurones. This compound, 2-hydroxy-saclofen, may be useful in assessing the physiological significance of central baclofen receptors.
Sigma-Aldrich
2-Hydroxysaclofen, ≥98% (TLC), solid