Merck
  • A novel glucose sensor using lutetium phthalocyanine as redox mediator in reduced graphene oxide conducting polymer multifunctional hydrogel.
A novel glucose sensor using lutetium phthalocyanine as redox mediator in reduced graphene oxide conducting polymer multifunctional hydrogel.
Biosensors & bioelectronics (2016-11-12)
H Al-Sagur, S Komathi, M A Khan, A G Gurek, A Hassan
ABSTRACT
Herein, we report a scalable synthesis of multifunctional conducting polyacrylic acid (PAA) hydrogel (MFH) integrated with reduced grapheme oxide (rGO), vinyl substituted polyaniline (VS-PANI) and lutetium Phthalocyanine (LuPc
Sigma-Aldrich
3-Vinylaniline, 97%
Pricing and availability is not currently available.