Merck
  • Inhibition of the glutamate transporter and glial enzymes in rat striatum by the gliotoxin, alpha aminoadipate.
Inhibition of the glutamate transporter and glial enzymes in rat striatum by the gliotoxin, alpha aminoadipate.
British journal of pharmacology (1994-10-01)
G J McBean
ABSTRACT
1. The effect of the gliotoxic analogue of glutamate, alpha aminoadipate, on the high affinity transport of D-[3H]-aspartate into a crude striatal P2 preparation, and on the activity of two enzymes of which glutamate is the substrate has been examined. 2. The L-isomer of alpha aminoadipate competitively inhibited the transport protein, with a Ki value of 192 microM, whereas the D-isomer of alpha aminoadipate was ineffective. The potent convulsant, L-methionine-S-sulphoximine, was also without effect on the activity of the glutamate transport protein. 3. L-alpha Aminoadipate was a competitive inhibitor of both glutamine synthetase, and gamma-glutamylcysteine synthetase, with Ki values of 209 microM and 7 mM respectively. Once again, the D-isomer of alpha aminoadipate was a far weaker inhibitor of either enzyme. 4. The results are discussed in terms of the mechanism of action of alpha aminoadipate in causing toxicity of glial cells.
Sigma-Aldrich
DL-2-Aminoadipic acid, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
D-2-Aminoadipic acid
Pricing and availability is not currently available.
Sigma-Aldrich
L-2-Aminoadipic acid, ≥98% (TLC)
Pricing and availability is not currently available.