Merck
  • Surface charge-bias impact of amine-contained pseudozwitterionic biointerfaces on the human blood compatibility.
Surface charge-bias impact of amine-contained pseudozwitterionic biointerfaces on the human blood compatibility.
Colloids and surfaces. B, Biointerfaces (2017-01-08)
Antoine Venault, Ko-Jen Hsu, Lu-Chen Yeh, Arunachalam Chinnathambi, Hsin-Tsung Ho, Yung Chang
ABSTRACT
This work discusses the impact of the charge bias and the hydrophilicity on the human blood compatibility of pseudozwitterionic biomaterial gels. Four series of hydrogels were prepared, all containing negatively-charged 3-sulfopropyl methacrylate (SA), and either acrylamide, N-isopropylacrylamide, 2-dimethylaminoethyl methacrylate (DMAEMA) or [2-(methacryloyloxy)ethyl]trimethylammonium (TMA), to form S
Sigma-Aldrich
3-Sulfopropyl methacrylate potassium salt, 98%