- Silica-supported Au@hollow-SiO
Silica-supported Au@hollow-SiO
Journal of colloid and interface science (2017-01-01)
Aruni Shajkumar, Bhanu Nandan, Sunita Sanwaria, Victoria Albrecht, Marcin Libera, Myong-Hoon Lee, Gudrun Auffermann, Manfred Stamm, Andriy Horechyy
PMID28039806
ABSTRACT
Catalytically active Au@hollow-SiO