Merck
  • Betaine attenuates memory impairment after water-immersion restraint stress and is regulated by the GABAergic neuronal system in the hippocampus.
Betaine attenuates memory impairment after water-immersion restraint stress and is regulated by the GABAergic neuronal system in the hippocampus.
European journal of pharmacology (2016-12-13)
Kazuo Kunisawa, Kiwamu Kido, Natsuki Nakashima, Takuya Matsukura, Toshitaka Nabeshima, Masayuki Hiramatsu
ABSTRACT
GABA mediated neuronal system regulates hippocampus-dependent memory and stress responses by controlling plasticity and neuronal excitability. Here, we demonstrate that betaine ameliorates water-immersion restraint stress (WIRS)-induced memory impairments. This improvement was inhibited by a betaine/GABA transporter-1 (GABA transporter-2: GAT2) inhibitor, NNC 05-2090. In this study, we investigated whether memory amelioration by betaine was mediated by the GABAergic neuronal system. Adult male mice were co-administered betaine and GABA receptor antagonists after WIRS. We also examined whether memory impairment after WIRS was attenuated by GABA receptor agonists. The memory functions were evaluated using a novel object recognition test 3-6 days after WIRS and/or the step-down type passive avoidance test at 7-8 days. The co-administration of the GABA
Sigma-Aldrich
Phaclofen, solid
Pricing and availability is not currently available.