Merck
Transcriptomic and anatomic parcellation of 5-HT
Nature communications (2017-01-31)
Sarah Frazer, Julien Prados, Mathieu Niquille, Christelle Cadilhac, Foivos Markopoulos, Lucia Gomez, Ugo Tomasello, Ludovic Telley, Anthony Holtmaat, Denis Jabaudon, Alexandre Dayer
ABSTRACT
Cortical GABAergic interneurons constitute a highly diverse population of inhibitory neurons that are key regulators of cortical microcircuit function. An important and heterogeneous group of cortical interneurons specifically expresses the serotonin receptor 3A (5-HT
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
Monoclonal Anti-MEIS2 antibody produced in mouse, clone 1H4, purified immunoglobulin, buffered aqueous solution