- 1-Methyl-tryptophan attenuates regulatory T cells differentiation due to the inhibition of estrogen-IDO1-MRC2 axis in endometriosis.
1-Methyl-tryptophan attenuates regulatory T cells differentiation due to the inhibition of estrogen-IDO1-MRC2 axis in endometriosis.
Cell death & disease (2016-12-03)
Chunyan Wei, Jie Mei, Lingli Tang, Yukai Liu, Dajin Li, Mingqing Li, Xiaoyong Zhu
PMID27906184
ABSTRACT
Foxp3