- Efficacy of periimplant mechanical debridement with and without adjunct antimicrobial photodynamic therapy in the treatment of periimplant diseases among cigarette smokers and non-smokers.
Efficacy of periimplant mechanical debridement with and without adjunct antimicrobial photodynamic therapy in the treatment of periimplant diseases among cigarette smokers and non-smokers.
The aim was to assess the efficacy of mechanical debridement (MD) with and without adjunct antimicrobial photodynamic therapy (aPDT) in reducing periimplant inflammation among cigarette-smokers and non-smokers. Cigarette-smokers and non-smokers were randomly divided into 2 groups. In the test-group, participants underwent full mouth scaling and periimplant MD with adjunct aPDT; and in the control-group, the participants underwent full mouth scaling and periimplant MD alone. Periimplant bleeding on probing (BOP), probing depth (PD) and crestal bone loss (CBL) were measured at baseline and at 6- and 12-months follow-up. Statistical analysis was performed using the Kruskal-Wallis test. P-values<0.05 were considered statistically significant. Eighty-four smokers (41 patients in the test group and 43 in the control group) and 82 non-smokers (40 patients in the test group and 42 in the control group) were included. Among smokers and non-smokers, periimplant PD was significantly higher in the control-group compared with the test-group (P<0.05) at 6-months of follow-up. There was no statistically significant difference in BOP, PD and CBL among smokers and non-smokers in the test- and control-groups at 12-months of follow-up. BOP was comparable among smokers at all time intervals. In the short-term, MD with adjunct aPDT is more effective in reducing periimplant probing depth than MD alone in smokers and non-smokers. However, in the long-term outcomes of MD either with or without aPDT among smokers and non-smokers are comparable.