- Miniature surface-plasmon resonance immunosensors--rapid and repetitive procedure.
Miniature surface-plasmon resonance immunosensors--rapid and repetitive procedure.
Analytical and bioanalytical chemistry (2002-04-09)
Masayasu Suzuki, Fumihiko Ozawa, Wakako Sugimoto, Shuji Aso
PMID11936102
ABSTRACT
Miniaturized immunosensors based on surface-plasmon resonance (SPR) have been developed by using the sensor recently developed by Texas Instruments. By using this sensor for human immunoglobulin G (IgG) rapid and repetitive measurements could be performed by alternate injection of sample and glycine-HCl buffer. A sensor for human serum albumin (HSA) could also be developed similarly. One difficulty in immunosensor development is dissociation of the strongly bound antigen-antibody complex. A "regenerable" immunosensor has also been developed for this reason.