Merck
  • Circadian RNA expression elicited by 3'-UTR IRAlu-paraspeckle associated elements.
Circadian RNA expression elicited by 3'-UTR IRAlu-paraspeckle associated elements.
eLife (2016-07-22)
Manon Torres, Denis Becquet, Marie-Pierre Blanchard, Séverine Guillen, Bénédicte Boyer, Mathias Moreno, Jean-Louis Franc, Anne-Marie François-Bellan
ABSTRACT
Paraspeckles are nuclear bodies form around the long non-coding RNA, Neat1, and RNA-binding proteins. While their role is not fully understood, they are believed to control gene expression at a post-transcriptional level by means of the nuclear retention of mRNA containing in their 3'-UTR inverted repeats of Alu sequences (IRAlu). In this study, we found that, in pituitary cells, all components of paraspeckles including four major proteins and Neat1 displayed a circadian expression pattern. Furthermore the insertion of IRAlu at the 3'-UTR of the EGFP cDNA led to a rhythmic circadian nuclear retention of the egfp mRNA that was lost when paraspeckles were disrupted whereas insertion of a single antisense Alu had only a weak effect. Using real-time video-microscopy, these IRAlu were further shown to drive a circadian expression of EGFP protein. This study shows that paraspeckles, thanks to their circadian expression, control circadian gene expression at a post-transcriptional level.
Sigma-Aldrich
Anti-PSPC1 (N-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-PSPC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.