Merck
  • Corticosteroids induce cyclooxygenase 1 expression in cardiomyocytes: role of glucocorticoid receptor and Sp3 transcription factor.
Corticosteroids induce cyclooxygenase 1 expression in cardiomyocytes: role of glucocorticoid receptor and Sp3 transcription factor.
Molecular endocrinology (Baltimore, Md.) (2008-07-05)
Haipeng Sun, Elena Sheveleva, Qin M Chen
ABSTRACT
Cyclooxygenase (COX) encodes a rate-limiting enzyme in the biosynthesis of prostanoids. Although COX-1 is constitutively expressed in many tissues, we found that glucocorticoids cause elevated expression of COX-1 gene in cardiomyocytes. Corticosterone (CT) at physiologically relevant doses (0.05-1 microm) induces transcriptional activation of COX-1 gene as shown by nuclear run-on and promoter reporter assays. An antagonist of glucocorticoid receptor (GR), mifepristone, prevented CT from inducing COX-1. COX-1 gene promoter deletion and mutation studies indicate a role of Sp transcription factors in CT-induced COX-1 gene. EMSAs or chromatin immunoprecipitation assays suggest that GR and Sp3 transcription factor bind to the promoter of COX-1 gene. Coimmunoprecipitation assays found an association of GR with Sp3. Silencing Sp3 protein with small interfering RNA suppressed CT-induced COX-1 promoter activation. Our data suggest that activated GR interacts with Sp3 transcription factor in binding to COX-1 promoter to enhance COX-1 gene expression in cardiomyocytes.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Anti-Sp1 Antibody, Upstate®, from rabbit