Merck
  • Loss of the RNA-binding protein TACO1 causes late-onset mitochondrial dysfunction in mice.
Loss of the RNA-binding protein TACO1 causes late-onset mitochondrial dysfunction in mice.
Nature communications (2016-06-21)
Tara R Richman, Henrik Spåhr, Judith A Ermer, Stefan M K Davies, Helena M Viola, Kristyn A Bates, John Papadimitriou, Livia C Hool, Jennifer Rodger, Nils-Göran Larsson, Oliver Rackham, Aleksandra Filipovska
ABSTRACT
The recognition and translation of mammalian mitochondrial mRNAs are poorly understood. To gain further insights into these processes in vivo, we characterized mice with a missense mutation that causes loss of the translational activator of cytochrome oxidase subunit I (TACO1). We report that TACO1 is not required for embryonic survival, although the mutant mice have substantially reduced COXI protein, causing an isolated complex IV deficiency. We show that TACO1 specifically binds the mt-Co1 mRNA and is required for translation of COXI through its association with the mitochondrial ribosome. We determined the atomic structure of TACO1, revealing three domains in the shape of a hook with a tunnel between domains 1 and 3. Mutations in the positively charged domain 1 reduce RNA binding by TACO1. The Taco1 mutant mice develop a late-onset visual impairment, motor dysfunction and cardiac hypertrophy and thus provide a useful model for future treatment trials for mitochondrial disease.
Sigma-Aldrich
Anti-MRPS34 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.