Merck
  • Long-term regulation of N-methyl-D-aspartate receptor subunits and associated synaptic proteins following hippocampal synaptic plasticity.
Long-term regulation of N-methyl-D-aspartate receptor subunits and associated synaptic proteins following hippocampal synaptic plasticity.
Neuroscience (2003-05-07)
J M Williams, D Guévremont, J T T Kennard, S E Mason-Parker, W P Tate, W C Abraham
ABSTRACT
Synaptic plasticity in the dentate gyrus is dependent on activation of the N-methyl-D-aspartate (NMDA)-subtype of glutamate receptors. In this study, we show that synaptic plasticity in turn regulates NMDA receptors, since subunits of the NMDA receptor complex are bidirectionally and independently regulated in the dentate gyrus following activation of perforant synapses in awake animals. Low-frequency stimulation that produced a mild synaptic depression resulted in a decrease in the NMDA receptor subunits NR1 and NR2B 48 h following stimulation. High-frequency stimulation that produced long-term potentiation resulted in an increase in NR1 and NR2B at the same time point. Further investigations revealed that in contrast to NR2B, NR1 levels increased gradually after long-term potentiation induction, reaching a peak level at 48 h, and were insensitive to the competitive NMDA receptor antagonist 3-3(2-carboxypiperazin-4-yl) propyl-1-phosphate. The increased levels of NR1 and NR2B at 48 h were found associated with synaptic membranes and with increased NMDA receptor-associated proteins, postsynaptic density protein 95, neuronal nitric oxide synthase and Ca(2+)/calmodulin-dependent protein kinase II, alpha subunit. These data suggest that the persistence of long-term potentiation is associated with an increase in the number of NMDA receptor complexes, which may be indicative of an increase in synaptic contact area.
Product Description
Millipore
Nylon Membrane Filter, 100.0 μm Pore Size, Millipore, filter diam. 90 mm, hydrophilic