Merck
  • Per(3-chloro-4-methyl)phenylcarbamate cyclodextrin clicked stationary phase for chiral separation in multiple modes high-performance liquid chromatography.
Per(3-chloro-4-methyl)phenylcarbamate cyclodextrin clicked stationary phase for chiral separation in multiple modes high-performance liquid chromatography.
Analytica chimica acta (2016-11-09)
Jian Tang, Limin Pang, Jie Zhou, Shaopeng Zhang, Weihua Tang
ABSTRACT
In this work, a detail study has been performed on the enantioselectivity of per(3-chloro-4-methyl)phenylcarbamate-β-CD clicked chiral stationary phase (CSP) in high-performance liquid chromatography. Both normal phase and polar organic mobile phases have been explored for the enantioseparation of 39 model racemic pairs including aromatic alcohols, flavonoids, β-blockers and amino acids. Chiral resolution values over 10 were achieved for flavonoids. The comparison study with reference perphenylcarbamate-β-CD clicked CSP reveals the significance of 3-chloro-4-methyl functionality in improving the enantioselectivities. This study may provide insight on the multiple interactions between functionalized CD and analytes.