Merck
  • Extractive scintillating polymer sensors for trace-level detection of uranium in contaminated ground water.
Extractive scintillating polymer sensors for trace-level detection of uranium in contaminated ground water.
Analytica chimica acta (2016-11-17)
Christine E Duval, Timothy A DeVol, Scott M Husson
ABSTRACT
This contribution describes the synthesis of robust extractive scintillating resin and its use in a flow-cell detector for the direct detection of uranium in environmental waters. The base poly[(4-methyl styrene)-co-(4-vinylbenzyl chloride)-co-(divinylbenzene)-co-(2-(1-napthyl)-4-vinyl-5-phenyloxazole)] resin contains covalently bound fluorophores. Uranium-binding functionality was added to the resin by an Arbuzov reaction followed by hydrolysis via strong acid or trimethylsilyl bromide (TMSBr)-mediated methanolysis. The resin was characterized by Fourier-transform infrared spectroscopy and spectrofluorometry. Fluorophore degradation was observed in the resin hydrolyzed by strong acid, while the resin hydrolyzed by TMSBr-mediated methanolysis maintained luminosity and showed hydrogen bonding-induced Stokes' shift of ∼100 nm. The flow cell detection efficiency for uranium of the TMSBr-mediated methanolysis resin was evaluated at pH 4, 5 and 6 in DI water containing 500 Bq L
Sigma-Aldrich
Triethyl phosphite, 98%