Merck
  • Reaction kinetic analysis of the 3-hydroxypropionate/4-hydroxybutyrate CO
Reaction kinetic analysis of the 3-hydroxypropionate/4-hydroxybutyrate CO
Metabolic engineering (2016-10-25)
Andrew J Loder, Yejun Han, Aaron B Hawkins, Hong Lian, Gina L Lipscomb, Gerrit J Schut, Matthew W Keller, Michael W W Adams, Robert M Kelly
ABSTRACT
The 3-hydroxypropionate/4-hydroxybutyrate (3HP/4HB) cycle fixes CO
Sigma-Aldrich
Sodium acrylate, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Succinyl coenzyme A sodium salt, ≥85%