Merck
  • Treatment by serum up-conversion nanoparticles in the fluoride matrix changes the mechanism of cell death and the elasticity of the membrane.
Treatment by serum up-conversion nanoparticles in the fluoride matrix changes the mechanism of cell death and the elasticity of the membrane.
Micron (Oxford, England : 1993) (2016-08-28)
Svetlana N Pleskova, Ekaterina N Gorshkova, Viktor V Novikov, Marc Solioz
ABSTRACT
Nanoparticles are increasingly being used for treatment and diagnostic purposes, but their effects on cells is not fully understood. Here, the interaction of fluorescent up-conversion nanoparticles (UpC-NPs) with neutrophils was investigated by imaging and measurement of membrane-cytosceletal elasticity by atomic force microscopy. It was found that UpC-NPs induce the death of neutrophils mainly by necrosis, and to a smaller extent by a novel process called 'mummification'. Necrosis occurs by gradual loss of intracellular contents and nuclei, 45-110min after exposure to UpC-NPs. Mummification is apparent as an increase in the rigidity of the neutrophils' membrane and acquisition of a characteristic bumpy shape with numerous protrusions; this structure does not change during atomic force microscopy scanning. Coating UpC-NPs with protein by incubation with serum leads to (1) formation of nanoparticle aggregates in the nm and μm size range, (2) a reduction in toxicity, (3) reduced mummification of neutrophils, and (4) no significant reduction of the elasticity of the membrane-cytoskeletal complex of neutrophils 30min after exposure to coated UpC-NPs. The study shows that serum proteins greatly curb the toxicity of nanoparticles and reveals mummification as a novel mechanism of UpC-NP-induced cell death.
Sigma-Aldrich
Ytterbium(III) fluoride, anhydrous, powder, 99.98% trace metals basis
Pricing and availability is not currently available.