Merck
  • Poly(I)-poly(C12U) but not ribavirin prevents death in a hamster model of Nipah virus infection.
Poly(I)-poly(C12U) but not ribavirin prevents death in a hamster model of Nipah virus infection.
Antimicrobial agents and chemotherapy (2006-04-28)
M C Georges-Courbot, H Contamin, C Faure, P Loth, S Baize, P Leyssen, J Neyts, V Deubel
ABSTRACT
Clinical nonrandomized trials demonstrate some efficacy for ribavirin in the treatment of patients with severe Nipah virus-induced encephalitis. We report here that EICAR, the 5-ethynyl analogue of ribavirin, and the OMP-decarboxylase inhibitors 6-aza-uridine and pyrazofurin have strong antiviral activity against Nipah virus replication in vitro. Ribavirin and 6-aza-uridine were tested further in hamsters infected with a lethal dose of Nipah virus. The activity of these small-molecule inhibitors was compared with that of the interferon inducer poly(I)-poly(C(12)U). Both ribavirin and 6-aza-uridine were able to delay but not prevent Nipah virus-induced mortality. Poly(I)-poly(C(12)U), at 3 mg/kg of body weight daily from the day of infection to 10 days postinfection, prevented mortality in 5 of 6 infected animals.
Sigma-Aldrich
Pyrazofurin, ≥98% (HPLC)
Pricing and availability is not currently available.