Merck
  • 6-(4-Benzylpiperazin-1-yl) benzodioxanes as selective ligands at cloned primate dopamine D 4 receptors.
6-(4-Benzylpiperazin-1-yl) benzodioxanes as selective ligands at cloned primate dopamine D 4 receptors.
Bioorganic & Medicinal Chemistry (2001)
Hodgetts KJ, et al.