Merck
  • Mild and general methods for the palladium-catalyzed cyanation of aryl and heteroaryl chlorides.
Mild and general methods for the palladium-catalyzed cyanation of aryl and heteroaryl chlorides.
Organic letters (2007-03-28)
Adam Littke, Maxime Soumeillant, Robert F Kaltenbach, Robert J Cherney, Christine M Tarby, Susanne Kiau
ABSTRACT
[reaction: see text] New methods for the palladium-catalyzed cyanation of aryl and heteroaryl chlorides have been developed, featuring sterically demanding, electron-rich phosphines. Highly challenging electron-rich aryl chlorides, in addition to electron-neutral and electron-deficient substrates, as well as nitrogen- and sulfur-containing heteroaryl chlorides can all undergo efficient cyanation under relatively mild conditions using readily available materials. In terms of substrate scope and temperature, these methods compare very favorably with the state-of-the-art cyanations of aryl chlorides.
Sigma-Aldrich
2-Amino-4-chloropyrimidine, 97%
Pricing and availability is not currently available.