- Identification of derivatives of bisphenol A diglycidyl ether and novolac glycidyl ether in can coatings by liquid chromatography/ion trap mass spectrometry.
Identification of derivatives of bisphenol A diglycidyl ether and novolac glycidyl ether in can coatings by liquid chromatography/ion trap mass spectrometry.
A reversed-phase liquid chromatographic method combined with fluorescence and multiple mass spectrometric detection in series is presented for the separation and structure elucidation of bisphenol A diglycidyl ether (BADGE) and novolac glycidyl ether (NOGE) derivatives. Atmospheric pressure chemical ionization in the positive ion mode and collision induced fragmentation in the ion trap allowed identification of BADGE- and NOGE-related compounds originating from reactions of the glycidyl ethers with bisphenols, solvents, and chain stoppers. Two extracts from food-can coatings were investigated in detail. It was possible to elucidate the structures of many substances and consequently to draw conclusions about the production of the lacquers.