Merck
  • Biaryl synthesis via Pd-catalyzed decarboxylative coupling of aromatic carboxylates with aryl halides.
Biaryl synthesis via Pd-catalyzed decarboxylative coupling of aromatic carboxylates with aryl halides.
Journal of the American Chemical Society (2007-03-23)
Lukas J Goossen, Nuria Rodríguez, Bettina Melzer, Christophe Linder, Guojun Deng, Laura M Levy
ABSTRACT
A new strategy for the regiospecific construction of unsymmetrical biaryls is presented, in which easily available salts of carboxylic acids are decarboxylated in situ to give arylmetal species that serve as the nucleophilic component in a catalytic cross-coupling reaction with aryl halides. The catalyst system consists of a copper phenanthroline complex that mediates the extrusion of CO2 from aromatic carboxylates to generate arylcopper species, and a palladium complex that catalyzes the cross-coupling of these intermediates with aryl halides. This bimetallic system allows the direct coupling of various aryl, heteroaryl, or vinyl carboxylic acids with aryl or heteroaryl iodides, bromides, or chlorides at 160 degrees C in the presence of a mild base such as potassium carbonate. The present scope and potential economic impact of the reaction are demonstrated by the synthesis of 42 biaryls, some of which are of substantial industrial relevance. Remaining challenges and future perspectives of the new transformation are discussed.
Sigma-Aldrich
Palladium(II) acetylacetonate, 99%
Pricing and availability is not currently available.