Merck
  • Observational and mechanistic links between C-reactive protein and blood pressure in elderly women.
Observational and mechanistic links between C-reactive protein and blood pressure in elderly women.
Maturitas (2016-05-18)
Adrian Hosford-Donovan, Andreas Nilsson, Britta Wåhlin-Larsson, Fawzi Kadi
ABSTRACT
It is hypothesized that chronic systemic inflammation contributes to the age-related decline in cardiovascular function. The aim of the present study was to combine an assessment of the relationship between the serum level of C-reactive protein (CRP) and systolic and diastolic blood pressure in 108 elderly women (65 and 70 years) with an in-vitro exploration of the effects of CRP on the proliferative and angiogenic potential of endothelial cells exposed to serum in elderly women. Based on the median CRP level in our population, LowCRP (CRP<1.3mg/L) and HighCRP (>1.3mg/L) groups were identified. Body mass index, waist circumference, systolic blood pressure (SBP) and diastolic blood pressure (DBP) were significantly higher in the HighCRP group than in the LowCRP group (p<0.05). The influence of CRP on SBP and DBP remained significant after adjustments for BMI and use of antihypertensive medication (p<0.05). When adjusting for waist circumference the observed influence of CRP on SPB was attenuated (p=0.062). We next evaluated the ability to form capillary tubes (angiogenesis assay) and the proliferation rate of endothelial cells exposed to the sera of elderly women. Increased serum CRP levels were associated with an increased doubling time of endothelial cells (R(2)=0.39; p<0.05) and decreased capillary tube length (R(2)=0.30; p<0.05), indicating a reduction in the proliferation rate of endothelial cells and angiogenic potential. In conclusion, chronic inflammation influences blood pressure in elderly women and compromises endothelial cell function, thus contributing to the age-related decline in vascular health.
Sigma-Aldrich
C Reactive Protein human, recombinant, expressed in E. coli, liquid
Pricing and availability is not currently available.