Merck
  • Vitamin D3 alters microglia immune activation by an IL-10 dependent SOCS3 mechanism.
Vitamin D3 alters microglia immune activation by an IL-10 dependent SOCS3 mechanism.
Journal of neuroimmunology (2016-03-06)
Mandy Boontanrart, Samuel D Hall, Justin A Spanier, Colleen E Hayes, Julie K Olson
ABSTRACT
Microglia become activated immune cells during infection or disease in the central nervous system (CNS). However, the mechanisms that downregulate activated microglia to prevent immune-mediated damage are not completely understood. Vitamin D3 has been suggested to have immunomodulatory affects, and high levels of vitamin D3 have been correlated with a decreased risk for developing some neurological diseases. Recent studies have demonstrated the synthesis of active vitamin D3, 1,25-dihydroxyvitamin D3, within the CNS, but its cellular source and neuroprotective actions remain unknown. Therefore, we wanted to determine whether microglia can respond to vitamin D3 and whether vitamin D3 alters immune activation of microglia. We have previously shown that microglia become activated by IFNγ or LPS or by infection with virus to express pro-inflammatory cytokines, chemokines, and effector molecules. In this study, activated microglia increased the expression of the vitamin D receptor and Cyp27b1, which encodes the enzyme for converting vitamin D3 into its active form, thereby enhancing their responsiveness to vitamin D3. Most importantly, the activated microglia exposed to vitamin D3 had reduced expression of pro-inflammatory cytokines, IL-6, IL-12, and TNFα, and increased expression of IL-10. The reduction in pro-inflammatory cytokines was dependent on IL-10 induction of suppressor of cytokine signaling-3 (SOCS3). Therefore, vitamin D3 increases the expression of IL-10 creating a feedback loop via SOCS3 that downregulates the pro-inflammatory immune response by activated microglia which would likewise prevent immune mediated damage in the CNS.
Sigma-Aldrich
MISSION® esiRNA, targeting human SOCS3
Pricing and availability is not currently available.