Merck
  • Distribution and Sources of Petroleum Hydrocarbons in Recent Sediments of the Imo River, SE Nigeria.
Distribution and Sources of Petroleum Hydrocarbons in Recent Sediments of the Imo River, SE Nigeria.
Archives of environmental contamination and toxicology (2015-11-08)
Inyang O Oyo-Ita, Orok E Oyo-Ita, Miranda I Dosunmu, Carmen Domínguez, Josep M Bayona, Joan Albaigés
ABSTRACT
The distribution of aliphatic and aromatic hydrocarbons in surface sediments of the lower course of the Imo River (Nigeria) was investigated to determine the sources and fate of these compounds. The aliphatic fraction is characterized by a widespread contribution of highly weathered/biodegraded hydrocarbon residues (reflected in the absence of prominent n-alkane peaks coupled with the presence of 17α(H),21β(H)-25-norhopane, an indicator of heavy hydrocarbon biodegradation) of Nigerian crude oils (confirmed by the occurrence of 18α(H)-oleanane, a compound characteristic of oils of deltaic origin). The concentrations of polycyclic aromatic hydrocarbons (PAHs) ranging from 48 to 117 ng/g dry weight (dw; ∑13PAHs) indicate a moderate pollution, possibly lowered by the sandy lithology and low organic carbon (OC) content of the sediments. Concentrations slightly decrease towards the estuary of the river, probably due to the fact that these stations are affected by tidal flushing of pollutants adsorbed on sediment particles and carried away by occasional storm to the Atlantic Ocean. A number of PAH ratios, including parent/alkylated and isomeric compounds, indicates a predominance of petrogenic sources, with a low contribution of pyrolytic inputs, particularly of fossil fuel combustion. On the basis of OC/ON (>10) and Per/ΣPAHpenta- (>10) values, a diagenetic terrigenous OC was proposed as a source of perylene to the river.
Sigma-Aldrich
Benzo[a]pyrene-d12, 98 atom % D
Sigma-Aldrich
Pyrene-d10, 98 atom % D
Sigma-Aldrich
Anthracene-d10, 98 atom % D