Merck
  • Towards the total chemical synthesis of integral membrane proteins: a general method for the synthesis of hydrophobic peptide-thioester building blocks.
Towards the total chemical synthesis of integral membrane proteins: a general method for the synthesis of hydrophobic peptide-thioester building blocks.
Tetrahedron letters (2007-03-05)
Erik C B Johnson, Stephen B H Kent
ABSTRACT
Modification of a peptide-(α)thioester with a sequence of six arginines on the thioester leaving group can render soluble all peptides derived from a polytopic integral membrane protein. This strategy greatly simplifies the synthesis of peptide-(α)thioester building blocks for the total chemical synthesis of integral membrane proteins by native chemical ligation.
Sigma-Aldrich
2-Amino-2-methyl-1-propanol, ~5% Water, technical grade, 95%
Pricing and availability is not currently available.