Merck
  • Balance and coordination training, but not endurance training, enhances synaptophysin and neurotrophin-3 immunoreactivity in the lumbar spinal cord after sciatic nerve crush.
Balance and coordination training, but not endurance training, enhances synaptophysin and neurotrophin-3 immunoreactivity in the lumbar spinal cord after sciatic nerve crush.
Muscle & nerve (2015-09-01)
Leandro Viçosa Bonetti, Jocemar Ilha, Ana Paula Krauthein Schneider, Silvia Barbosa, Maria Cristina Faccioni-Heuser
ABSTRACT
Numerous rehabilitation treatments have been shown to be useful for peripheral and central restoration after (PNI). After sciatic nerve crush, we investigated 4 weeks of endurance training (ET) and balance and coordination training (BCT) with sciatic function index, hind-paw stride length, and spinal cord dorsal horn synaptophysin and neurotrophin-3 immunoreactivity. Our results demonstrated no significant differences between the non-trained (NT), ET, and BCT groups in sciatic functional index, and in stride-length analysis, but the ET showed higher values compared with the NT group. Synaptophysin immunoreactivity was higher in the BCT group compared with the NT group, and neurotrophin-3 immunoreactivity in the BCT group was greater compared with the other groups. BCT can positively affect spinal cord plasticity after a (PNI), and these modifications are important in the rehabilitation process.
Sigma-Aldrich
Anti-Neurotrophin 3 Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Triton X-100, laboratory grade
Pricing and availability is not currently available.