Merck
  • Synthesis of protected chiral vicinal diaminoalcohols by diastereoselective intramolecular benzylic substitution from bistrichloroacetimidates.
Synthesis of protected chiral vicinal diaminoalcohols by diastereoselective intramolecular benzylic substitution from bistrichloroacetimidates.
Organic letters (2007-01-16)
Christophe Rondot, Pascal Retailleau, Jieping Zhu
ABSTRACT
An efficient synthesis of chiral dihydrooxazines (2) from 1-aryl-2-amino-propane-1,3-diols (1) via the corresponding bistrichloroacetimidate intermediates has been developed. In this transformation, one trichloroacetimidate acts as a leaving group and the other acts as a nucleophile. The cyclization proceeds through an SN1 mechanism to provide trans-dihydrooxazines with complete diastereoselectivity irrespective of the absolute configuration of the benzylic alcohol. The transformation of 2 into other selectively protected aminodiols is also documented. [reaction: see text].
Sigma-Aldrich
Trichloroacetonitrile, 98%
Pricing and availability is not currently available.