Merck
  • Enhanced dihydropyridine receptor calcium channel activity restores muscle strength in JP45/CASQ1 double knockout mice.
Enhanced dihydropyridine receptor calcium channel activity restores muscle strength in JP45/CASQ1 double knockout mice.
Nature communications (2013-02-28)
Barbara Mosca, Osvaldo Delbono, Maria Laura Messi, Leda Bergamelli, Zhong-Min Wang, Mirko Vukcevic, Ruben Lopez, Susan Treves, Miyuki Nishi, Hiroshi Takeshima, Cecilia Paolini, Marta Martini, Giorgio Rispoli, Feliciano Protasi, Francesco Zorzato
ABSTRACT
Muscle strength declines with age in part due to a decline of Ca(2+) release from sarcoplasmic reticulum calcium stores. Skeletal muscle dihydropyridine receptors (Ca(v)1.1) initiate muscle contraction by activating ryanodine receptors in the sarcoplasmic reticulum. Ca(v)1.1 channel activity is enhanced by a retrograde stimulatory signal delivered by the ryanodine receptor. JP45 is a membrane protein interacting with Ca(v)1.1 and the sarcoplasmic reticulum Ca(2+) storage protein calsequestrin (CASQ1). Here we show that JP45 and CASQ1 strengthen skeletal muscle contraction by modulating Ca(v)1.1 channel activity. Using muscle fibres from JP45 and CASQ1 double knockout mice, we demonstrate that Ca(2+) transients evoked by tetanic stimulation are the result of massive Ca(2+) influx due to enhanced Ca(v)1.1 channel activity, which restores muscle strength in JP45/CASQ1 double knockout mice. We envision that JP45 and CASQ1 may be candidate targets for the development of new therapeutic strategies against decay of skeletal muscle strength caused by a decrease in sarcoplasmic reticulum Ca(2+) content.
Product Description
Roche
cOmplete ULTRA Tablets, EDTA-free, glass vials Protease Inhibitor Cocktail, Tablets provided in glass vials
Pricing and availability is not currently available.