Merck
  • Vasoconstrictive effect of angiotensin IV in isolated rat basilar artery independent of AT1 and AT2 receptors.
Vasoconstrictive effect of angiotensin IV in isolated rat basilar artery independent of AT1 and AT2 receptors.
Journal of vascular research (2005-10-29)
Sebastien Faure, James Javellaud, Jean-Michel Achard, Nicole Oudart
ABSTRACT
The effect of angiotensin IV (AngIV) was studied in freshly isolated rat basilar arteries (BAs) perfused at a constant rate. AngIV had no effect on basal BA perfusion pressure, but induced a marked concentration-dependent contraction in vessels precontracted by a 50-mM KCl solution (EC50=44.5+/-16 nM). This contraction was unaffected by the angiotensin AT1 receptor antagonist candesartan or the angiotensin AT2 receptor blocker PD123319, but was markedly inhibited by two different specific AT4 receptor antagonists, Nle1-Leu3 yen(CH2-NH2)3-4-AngIV and divalinal-AngIV. Removal of the endothelium abolished the contractile response to AngIV, and pretreatment of endothelium-intact arteries with the endothelin ETA/ETB receptors inhibitor PD142893 blocked the AngIV-induced contraction to the same extent. In BA pretreated with endothelin-1 (ET-1; 0.01 microM), AngIV-induced a concentration-dependent contraction, shifted to the left, compared with that observed with KCl precontraction, unaffected by candesartan but completely abolished by Nle1-Leu3 yen(CH2-NH2)3-4-AngIV. The contractile effect was not affected by endothelium removal in the presence of exogenous ET-1, in contrast to KCl pretreated BA, suggesting that endothelium was mandatory to unmask the effect of AngIV as a source of endogenous ET-1 release. Taken together, these results indicate that low (nanomolar) concentrations of AngIV exert a constrictive effect mediated by its specific binding site AT4 in the rat BA, and that this vasoactive effect is indirect and involves endogenous endothelin(s).
Sigma-Aldrich
Endothelin Antagonist PD 142893
Pricing and availability is not currently available.