Merck
  • Clinical significance of P-glycoprotein and glutathione S-transferase π expression in gallbladder carcinoma.
Clinical significance of P-glycoprotein and glutathione S-transferase π expression in gallbladder carcinoma.
Experimental and therapeutic medicine (2014-02-13)
Qing-Jiu Ma, Yu-Cun Zhang, Jing-Sen Shi, Guo-Cai Li
ABSTRACT
P-glycoprotein (P-gp) and glutathione S-transferase π (GST-π) are not only drug-resistance markers, but also prognostic markers of various cancers. The aim of the present study was to investigate the clinical significance of P-gp and GST-π in gallbladder carcinoma (GBC). Tissue samples from 42 patients with GBC were immunostained. Demographic, clinical and follow-up data were collected and analyzed. The positive expression rates of P-gp and GST-π in the GBC tissues were significantly higher (76.2 and 64.3%, respectively) than that of chronic cholecystitis specimens (30 and 20%, respectively) (P=0.014 and 0.035, respectively), and correlated with the Nevin stage of GBC. Multivariate analysis demonstrated that patients with positive expression of P-gp and GST-π showed a significantly lower 2-year survival rate (11.1 and 12%, respectively) compared with patients with negative expression (55.6 and 45.5%, respectively) (P=0.013 and 0.036, respectively). P-gp was also found to be an independent prognostic marker of 2-year survival rate by logistic regression analysis (B=-2.76, P=0.061). Results of this study suggest that P-gp is a prognostic marker of GBC and the detection of P-gp and GST-π may contribute to the prognosis of GBC and the application of chemotherapy as a therapeutic treatment.
Sigma-Aldrich
Monoclonal Anti-P-Glycoprotein (MDR) antibody produced in mouse, clone F4, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution