Merck
  • Argonaute 2 sustains the gene expression program driving human monocytic differentiation of acute myeloid leukemia cells.
Argonaute 2 sustains the gene expression program driving human monocytic differentiation of acute myeloid leukemia cells.
Cell death & disease (2013-11-23)
I Iosue, R Quaranta, S Masciarelli, G Fontemaggi, E M Batassa, C Bertolami, T Ottone, M Divona, B Salvatori, F Padula, A Fatica, F Lo-Coco, C Nervi, F Fazi
ABSTRACT
MicroRNAs are key regulators of many biological processes, including cell differentiation. These small RNAs exert their function assembled in the RNA-induced silencing complexes (RISCs), where members of Argonaute (Ago) family of proteins provide a unique platform for target recognition and gene silencing. Here, by using myeloid cell lines and primary blasts, we show that Ago2 has a key role in human monocytic cell fate determination and in LPS-induced inflammatory response of 1,25-dihydroxyvitamin D3 (D3)-treated myeloid cells. The silencing of Ago2 impairs the D3-dependent miR-17-5p/20a/106a, miR-125b and miR-155 downregulation, the accumulation of their translational targets AML1, VDR and C/EBPβ and monocytic cell differentiation. Moreover, we show that Ago2 is recruited on miR-155 host gene promoter and on the upstream region of an overlapping antisense lncRNA, determining their epigenetic silencing, and miR-155 downregulation. These findings highlight Ago2 as a new factor in myeloid cell fate determination in acute myeloid leukemia cells.
Sigma-Aldrich
RIPAb+ Ago2 - RIP Validated Antibody and Primer Set, ascites fluid, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Pricing and availability is not currently available.