Merck
  • Sorting of GPI-anchored proteins into ER exit sites by p24 proteins is dependent on remodeled GPI.
Sorting of GPI-anchored proteins into ER exit sites by p24 proteins is dependent on remodeled GPI.
The Journal of cell biology (2011-07-06)
Morihisa Fujita, Reika Watanabe, Nina Jaensch, Maria Romanova-Michaelides, Tadashi Satoh, Masaki Kato, Howard Riezman, Yoshiki Yamaguchi, Yusuke Maeda, Taroh Kinoshita
ABSTRACT
Glycosylphosphatidylinositol (GPI) anchoring of proteins is a posttranslational modification occurring in the endoplasmic reticulum (ER). After GPI attachment, proteins are transported by coat protein complex II (COPII)-coated vesicles from the ER. Because GPI-anchored proteins (GPI-APs) are localized in the lumen, they cannot interact with cytosolic COPII components directly. Receptors that link GPI-APs to COPII are thought to be involved in efficient packaging of GPI-APs into vesicles; however, mechanisms of GPI-AP sorting are not well understood. Here we describe two remodeling reactions for GPI anchors, mediated by PGAP1 and PGAP5, which were required for sorting of GPI-APs to ER exit sites. The p24 family of proteins recognized the remodeled GPI-APs and sorted them into COPII vesicles. Association of p24 proteins with GPI-APs was pH dependent, which suggests that they bind in the ER and dissociate in post-ER acidic compartments. Our results indicate that p24 complexes act as cargo receptors for correctly remodeled GPI-APs to be sorted into COPII vesicles.
Sigma-Aldrich
Anti-ERGIC-53 antibody, Mouse monoclonal, clone ERGIC-3, purified from hybridoma cell culture